Assay Details
Target Gene Details
Entrez Gene ID: | 12292 |
Gene Name: | calcium channel, voltage-dependent, L type, alpha 1S subunit |
Gene Aliases: |
AW493108, Cav1.1, Cchl1a3, fmd, mdg, sj |
Location: |
Chr.1:136052805-136119822 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacna1s | NM_001081023.1 | NP_001074492.1 | ||
| NM_014193.2 | NP_055008.2 | |||
| AK142239.1 | BAE24989.1 | |||
| AK143541.1 | BAE25427.1 | |||
| BC134369.1 | ||||
| L06234.1 | AAB59700.1 |
More Information
Set Membership: |
|

Genomic Map