Assay Details
Target Gene Details
Entrez Gene ID: | 63913 |
Gene Name: | family with sequence similarity 129, member A |
Gene Aliases: |
AI256368, AU019833, Niban |
Location: |
Chr.1:151571373-151721939 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam129a | NM_022018.3 | 1 | 101 | NP_071301.2 |
| AK136597.1 | BAE23066.1 | |||
| AK170193.1 | BAE41626.1 | |||
| BC021332.1 | ||||
| BY016241.1 | 1 | 102 | ||
| BY194932.1 | 1 | 52 |

Genomic Map