Assay Details
Target Gene Details
Entrez Gene ID: | 240843 |
Gene Name: | bone morphogenic protein/retinoic acid inducible neural-specific 2 |
Gene Aliases: |
6430517E21Rik, BB361039, Fam5b, mKIAA1747 |
Location: |
Chr.1:158245265-158356528 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Brinp2 | NM_207583.2 | 6 | 1720 | NP_997466.2 |
| AK032297.1 | 6 | 1724 | ||
| AK045801.1 | 6 | 1247 | ||
| AK122549.1 | 6 | 1786 | BAC65831.1 | |
| BC076572.1 | 6 | 1720 | AAH76572.1 |

Genomic Map