Assay Details
Target Gene Details
Entrez Gene ID: | 214498 |
Gene Name: | cell division cycle 73, Paf1/RNA polymerase II complex component |
Gene Aliases: |
8430414L16Rik, BC027756, C130030P16Rik, C81219, Hrpt2 |
Location: |
Chr.1:143607499-143702888 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc73 | NM_145991.1 | 7 | 719 | NP_666103.1 |
| AK080861.1 | 7 | 851 | BAC38048.1 | |
| BC027756.1 | 7 | 820 | AAH27756.1 | |
| BC031127.1 | 7 | 719 | AAH31127.1 |

Genomic Map