Assay Details
Target Gene Details
Entrez Gene ID: | 108000 |
Gene Name: | centromere protein F |
Gene Aliases: |
6530404A22Rik, AI325968, CENF, Lek1, PRO1779, mitosin |
Location: |
Chr.1:189640606-189688105 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cenpf | NM_001081363.2 | 7 | 1134 | NP_001074832.2 |
| AK029517.1 | 7 | 1085 | BAC26488.1 | |
| AK080758.1 | BAC38010.2 | |||
| DQ642022.1 | 6 | 991 |

Genomic Map