Assay Details
Target Gene Details
Entrez Gene ID: | 14194 |
Gene Name: | fumarate hydratase 1 |
Gene Aliases: |
Fh, Fh-1 |
Location: |
Chr.1:175601378-175625635 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fh1 | NM_010209.2 | 7 | 1106 | NP_034339.2 |
| AK002379.1 | 7 | 1058 | ||
| AK147000.1 | 7 | 1100 | BAE27597.1 | |
| BC006048.1 | 7 | 1049 | AAH06048.1 |
More Information
Set Membership: |
|

Genomic Map