Assay Details
Target Gene Details
Entrez Gene ID: | 11545 |
Gene Name: | poly (ADP-ribose) polymerase family, member 1 |
Gene Aliases: |
5830444G22Rik, AI893648, ARTD1, Adprp, Adprt1, C80510, PARP, PPOL, parp-1, sPARP-1 |
Location: |
Chr.1:180568975-180601254 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Parp1 | NM_007415.2 | 10 | 1528 | NP_031441.2 |
| AF126717.1 | 1 | 161 | AAF61293.1 | |
| AK028042.1 | 10 | 1547 | ||
| AK030889.1 | 10 | 1531 | BAC27173.1 | |
| AK088679.1 | 10 | 1532 | BAC40500.1 | |
| AK146677.1 | 10 | 1549 | BAE27352.1 | |
| AK152360.1 | 10 | 1549 | BAE31150.1 | |
| AK159433.1 | 10 | 1580 | BAE35080.1 | |
| AK165158.1 | 10 | 1548 | ||
| AK166104.1 | 10 | 1580 | BAE38574.1 | |
| AK167699.1 | 10 | 1520 | BAE39743.1 | |
| AK169326.1 | 10 | 1549 | BAE41080.1 | |
| BC012041.1 | 10 | 1528 | AAH12041.1 | |
| X14206.1 | 10 | 1550 | CAA32421.1 |
More Information
Set Membership: |
|

Genomic Map