Assay Details
Target Gene Details
Entrez Gene ID: | 241113 |
Gene Name: | protein kinase, AMP-activated, gamma 3 non-catatlytic subunit |
Gene Aliases: |
AMPKg3L, AMPKg3S, Amkg3, Ampkg3 |
Location: |
Chr.1:74738922-74749590 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkag3 | NM_153744.3 | 13 | 1541 | NP_714966.1 |
| AF525500.1 | 13 | 1448 | AAN47137.1 | |
| AF525501.1 | 12 | 1469 | AAN47138.1 | |
| AK036585.1 | 13 | 1542 | BAC29492.1 | |
| BC116749.1 | 12 | 1405 | ||
| BC116777.1 | 12 | 1405 |
More Information
Set Membership: |
|

Genomic Map