Assay Details
Target Gene Details
Entrez Gene ID: | 74013 |
Gene Name: | raftlin family member 2 |
Gene Aliases: |
2700010E02Rik, 3222401M22Rik |
Location: |
Chr.1:55170159-55227065 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rftn2 | NM_028713.1 | 5 | 1383 | NP_082989.1 |
| AK012229.2 | 3 | 1078 | ||
| AK014318.1 | 5 | 1135 | BAB29269.1 | |
| BC026871.1 | 5 | 947 | AAH26871.1 | |
| BC038341.1 | 5 | 1383 | AAH38341.1 |

Genomic Map