Assay Details
Target Gene Details
Entrez Gene ID: | 269181 |
Gene Name: | mannoside acetylglucosaminyltransferase 4, isoenzyme A |
Gene Aliases: |
9530018I07Rik, GnT-IVa, glcNAc-T-IVa |
Location: |
Chr.1:37439340-37541047 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mgat4a | NM_001290801.1 | 1 | 217 | NP_001277730.1 |
| NM_173870.3 | 2 | 269 | NP_776295.1 | |
| AK082702.1 | 1 | 64 | ||
| AK155724.1 | 2 | 269 | ||
| AK163327.1 | 1 | 49 | BAE37301.1 |
More Information
Set Membership: |
|

Genomic Map