Assay Details
Target Gene Details
Entrez Gene ID: | 23956 |
Gene Name: | neuraminidase 2 |
Gene Aliases: |
MBS, MSS, MTS |
Location: |
Chr.1:87574027-87597845 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Neu2 | NM_001160163.1 | NP_001153635.1 | ||
| NM_001160164.1 | NP_001153636.1 | |||
| NM_001160165.1 | NP_001153637.1 | |||
| AB048604.1 | BAB39152.1 | |||
| AF139059.1 | ||||
| AK145284.1 | BAE26342.1 | |||
| BY088515.1 | ||||
| FJ613504.1 |
More Information
Set Membership: |
|

Genomic Map