Assay Details
Target Gene Details
Entrez Gene ID: | 545428 |
Gene Name: | coiled-coil domain containing 141 |
Gene Aliases: |
2610301F02Rik, CAMDI, ENSMUSG00000075261 |
Location: |
Chr.2:77009902-77176062 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc141 | NM_001025576.3 | NP_001020747.2 | ||
| AB540232.1 | ||||
| AK011952.1 | BAB27936.1 | |||
| AK041121.1 | BAC30830.1 | |||
| AK045498.1 | BAC32396.1 | |||
| AK053175.1 |

Genomic Map