Assay Details
Target Gene Details
Entrez Gene ID: | 545428 |
Gene Name: | coiled-coil domain containing 141 |
Gene Aliases: |
2610301F02Rik, CAMDI, ENSMUSG00000075261 |
Location: |
Chr.2:77009902-77176062 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc141 | NM_001025576.3 | 9 | 1488 | NP_001020747.2 |
| AB540232.1 | 9 | 1339 | ||
| AK041121.1 | 8 | 1295 | BAC30830.1 | |
| AK045498.1 | 9 | 1463 | BAC32396.1 | |
| AK053175.1 | 9 | 1474 |

Genomic Map