Assay Details
Target Gene Details
Entrez Gene ID: | 67693 |
Gene Name: | huntingtin interacting protein K |
Gene Aliases: |
2310003F16Rik, HSPC136 |
Location: |
Chr.2:121457088-121458440 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hypk | NM_026318.3 | NP_080594.2 | ||
| AI847440.1 | ||||
| AK003580.1 | BAB22870.2 | |||
| AK009109.1 | BAB26075.2 | |||
| AK168506.1 | BAE40389.1 | |||
| BC021588.1 | AAH21588.1 | |||
| BC038469.1 | AAH38469.1 | |||
| BY165803.1 |

Genomic Map