Assay Details
Target Gene Details
Entrez Gene ID: | 72931 |
Gene Name: | SWI5 recombination repair homolog (yeast) |
Gene Aliases: |
1500019F05Rik, 2900010J23Rik, AI046671 |
Location: |
Chr.2:32278805-32288136 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Swi5 | NM_001290552.1 | NP_001277481.1 | ||
| NM_175190.5 | NP_780399.1 | |||
| NR_110965.1 | ||||
| AB597964.1 | ||||
| AK013517.1 | BAC25409.1 | |||
| AK018775.1 | ||||
| AK031907.1 | ||||
| AK039840.1 | 2 | 215 | ||
| AK041567.1 | ||||
| AK088274.1 | ||||
| AK166608.1 | BAE38890.1 | |||
| BC021748.1 | AAH21748.1 |
More Information
Set Membership: |
|

Genomic Map