Assay Details
Target Gene Details
Entrez Gene ID: | 67135 |
Gene Name: | BPI fold containing family A, member 5 |
Gene Aliases: |
2310021H06Rik, 2310074B19Rik, Pluncl, Splunc5, Tpl |
Location: |
Chr.2:154162607-154168446 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bpifa5 | NM_025990.4 | NP_080266.2 | ||
| AK009329.1 | BAB26221.1 | |||
| AK009441.1 | BAB26290.1 | |||
| AK009531.1 | BAB26343.1 | |||
| AK009562.1 | BAB26360.1 | |||
| AK009580.1 | BAB26372.1 | |||
| AK009629.1 | BAB26401.1 | |||
| AK009695.1 | BAB26444.1 | |||
| AK009803.1 | BAB26513.1 | |||
| AK009835.1 | BAB26533.1 | |||
| AK010012.1 | BAB26642.1 | |||
| AK010051.1 | BAB26667.1 | |||
| AK010115.1 | BAB26710.1 | |||
| AK010143.1 | ||||
| AK010145.1 | BAB26730.1 | |||
| AK010158.1 | ||||
| AK010159.1 | ||||
| BC115526.1 | ||||
| BC115527.1 |

Genomic Map