Assay Details
Target Gene Details
Entrez Gene ID: | 73024 |
Gene Name: | ER membrane protein complex subunit 7 |
Gene Aliases: |
2900064A13Rik, AI451465, ORF3, c11orf3 |
Location: |
Chr.2:112455014-112472155 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Emc7 | NM_133749.2 | NP_598510.1 | ||
| AJ250345.1 | CAC16213.1 | |||
| AJ278128.1 | CAC01616.1 | |||
| AK013744.1 | ||||
| AK088738.1 | BAC40540.1 | |||
| AK201625.1 | ||||
| AK211388.1 | ||||
| BC008164.1 | AAH08164.1 | |||
| BC115477.1 | ||||
| BC115478.1 |

Genomic Map