Assay Details
Target Gene Details
Entrez Gene ID: | 27878 |
Gene Name: | transcriptional adaptor 1 |
Gene Aliases: |
2900026B15Rik, D1Ertd251e, Staf42, Tada1l |
Location: |
Chr.1:166379167-166393621 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tada1 | NM_030245.3 | 5 | 631 | NP_084521.1 |
| AK013599.1 | 5 | 398 | ||
| AK151132.1 | 5 | 493 | BAE30140.1 | |
| BC002307.1 | 5 | 433 | AAH02307.1 | |
| BC027337.1 | 5 | 498 | AAH27337.1 |

Genomic Map