Assay Details
Target Gene Details
Entrez Gene ID: | 21756 |
Gene Name: | protease, serine 40 |
Gene Aliases: |
Tesp2 |
Location: |
Chr.1:34536515-34560993 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss40 | NM_009356.2 | 1 | 106 | NP_033382.2 |
| AB008911.1 | 1 | 83 | BAA26133.1 | |
| AK132909.1 | 1 | 107 | BAE21416.1 | |
| BC145989.1 | 1 | 121 |
More Information
Set Membership: |
|

Genomic Map