Assay Details
Target Gene Details
Entrez Gene ID: | 228356 |
Gene Name: | RIKEN cDNA 1110051M20 gene |
Gene Aliases: |
AI586322 |
Location: |
Chr.2:91273240-91444713 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1110051M20Rik | NM_175123.4 | NP_780332.1 | ||
| AK018339.1 | ||||
| AK045977.1 | BAC32558.1 | |||
| AK164320.1 | BAE37736.1 | |||
| BC049648.1 | AAH49648.1 | |||
| BC085239.1 | AAH85239.1 | |||
| BC089006.1 | AAH89006.1 |

Genomic Map