Assay Details
Target Gene Details
Entrez Gene ID: | 67326 |
Gene Name: | RIKEN cDNA 1700037H04 gene |
Gene Aliases: |
- |
Location: |
Chr.2:131146315-131160086 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700037H04Rik | NM_026091.2 | NP_080367.1 | ||
| AK003406.1 | BAB22770.1 | |||
| AK006622.1 | ||||
| AK040365.1 | ||||
| AK048328.1 | 1 | 803 | ||
| AK178717.1 | ||||
| BC006041.1 | AAH06041.1 | |||
| BC071212.1 | AAH71212.1 |

Genomic Map