Assay Details
Target Gene Details
Entrez Gene ID: | 73489 |
Gene Name: | RIKEN cDNA 1700080N15 gene |
Gene Aliases: |
4933408D14Rik |
Location: |
Chr.2:4132065-4141141 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700080N15Rik | NR_040500.1 | |||
| AK006958.1 | ||||
| AK016735.1 |
Target Gene Details
Entrez Gene ID: | 209630 |
Gene Name: | FERM domain containing 4A |
Gene Aliases: |
2700017I06Rik, 5330420J21, AI594879, C230040M21Rik, Frmd4, Gm13190, OTTMUSG00000010806, mKIAA1294 |
Location: |
Chr.2:4017695-4614043 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Frmd4a |

Genomic Map