Assay Details
Target Gene Details
Entrez Gene ID: | 320706 |
Gene Name: | suppressor of glucose, autophagy associated 1 |
Gene Aliases: |
9830001H06Rik, AI426038, D430036N24Rik, Soga |
Location: |
Chr.2:157010442-157079265 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Soga1 | NM_001164663.1 | 3 | 1341 | NP_001158135.1 |
| AK147227.1 | 3 | 1332 | ||
| FJ977045.2 | 3 | 592 |

Genomic Map