Assay Details
Target Gene Details
Entrez Gene ID: | 241694 |
Gene Name: | Ral GTPase activating protein, alpha subunit 2 (catalytic) |
Gene Aliases: |
A230067G21Rik, AS250, BC053994, RGC2, pp250 |
Location: |
Chr.2:146239124-146512222 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ralgapa2 | NM_001033348.3 | NP_001028520.2 | ||
| CN706154.1 |
More Information
Set Membership: |
|

Genomic Map