Assay Details
Target Gene Details
Entrez Gene ID: | 77058 |
Gene Name: | coiled-coil domain containing 183 |
Gene Aliases: |
4921530D09Rik, Cccd183, Kiaa1984 |
Location: |
Chr.2:25608629-25617678 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc183 | NM_029859.1 | NP_084135.1 | ||
| AK014983.1 | BAB29653.1 |

Genomic Map