Assay Details
Target Gene Details
Entrez Gene ID: | 629499 |
Gene Name: | maestro heat-like repeat family member 8 |
Gene Aliases: |
4922505G16Rik |
Location: |
Chr.2:157208548-157279549 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mroh8 | NM_001039557.4 | NP_001034646.2 | ||
| AK029507.1 | ||||
| AK089157.1 | BAC40769.1 | |||
| AK132763.1 | BAE21344.1 | |||
| BC150937.1 |

Genomic Map