Assay Details
Target Gene Details
Entrez Gene ID: | 381356 |
Gene Name: | calcium channel flower domain containing 1 |
Gene Aliases: |
5930434B04Rik, mFwe |
Location: |
Chr.2:27009841-27021089 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacfd1 | NM_029862.4 | NP_084138.1 | ||
| AK039867.1 | BAC30468.1 | |||
| AK040379.1 | BAC30577.1 | |||
| AK052562.1 | BAC35040.1 | |||
| AK187500.1 |

Genomic Map