Assay Details
Target Gene Details
Entrez Gene ID: | 241688 |
Gene Name: | double zinc ribbon and ankyrin repeat domains 1 |
Gene Aliases: |
2810039F03Rik, 6330439K17Rik, 6430408C01, AI851115, Ankrd64 |
Location: |
Chr.2:144470555-144527425 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dzank1 | NM_172859.2 | 7 | 775 | NP_766447.2 |
| AK032182.1 | 7 | 775 | BAC27747.1 | |
| AK032632.1 | 7 | 766 | BAC27960.1 | |
| AK163994.1 | 7 | 766 | ||
| BC141156.1 | 7 | 674 | ||
| BC145464.1 | 7 | 674 |

Genomic Map