Assay Details
Target Gene Details
Entrez Gene ID: | 11464 |
Gene Name: | actin, alpha, cardiac muscle 1 |
Gene Aliases: |
Actc-1 |
Location: |
Chr.2:114047289-114052811 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Actc1 | NM_009608.3 | NP_033738.1 | ||
| AK014303.1 | BAB29258.1 | |||
| AK014307.1 | BAB29260.1 | |||
| AK146496.1 | ||||
| AK146899.1 | BAE27513.1 | |||
| AK168637.1 | BAE40497.1 | |||
| AK168661.1 | BAE40515.1 | |||
| AK168831.1 | BAE40656.1 | |||
| AK169161.1 | BAE40940.1 | |||
| AK169165.1 | BAE40944.1 | |||
| BC062138.1 | AAH62138.1 | |||
| BC100600.1 | AAI00601.1 | |||
| M15501.1 | AAA37167.1 | |||
| X03767.1 | CAA27398.1 |
More Information
Set Membership: |
|

Genomic Map