Assay Details
Target Gene Details
Entrez Gene ID: | 11480 |
Gene Name: | activin receptor IIA |
Gene Aliases: |
ActrIIa, Acvr2, TactrII |
Location: |
Chr.2:48814109-48903264 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acvr2a | NM_007396.4 | NP_031422.3 | ||
| AK041246.1 | BAC30877.1 | |||
| AK143186.1 | ||||
| BC138823.1 | ||||
| M65287.1 | AAA37171.1 |
More Information
Set Membership: |
|

Genomic Map