Assay Details
Target Gene Details
Entrez Gene ID: | 433470 |
Gene Name: | expressed sequence AA467197 |
Gene Aliases: |
Nmes1, mir-147 |
Location: |
Chr.2:122636971-122641087 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AA467197 | NM_001004174.1 | NP_001004174.1 | ||
| AK141297.1 | BAE24640.1 | |||
| AK150561.1 | BAE29660.1 | |||
| BC049633.1 | AAH49633.1 |
More Information
Set Membership: |
|

Genomic Map