Assay Details
Target Gene Details
Entrez Gene ID: | 68738 |
Gene Name: | acyl-CoA synthetase short-chain family member 1 |
Gene Aliases: |
1110032O15Rik, AI788978, Acas2, Acas2l, AceCS2 |
Location: |
Chr.2:150618111-150668240 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acss1 | NM_080575.2 | NP_542142.1 | ||
| AB046742.1 | BAB21612.1 | |||
| AK038517.1 | 1 | 665 | ||
| AK088244.1 | BAC40232.1 | |||
| AK146568.1 | BAE27266.1 | |||
| AK158119.1 | BAE34365.1 | |||
| AK172412.1 | BAE42994.1 | |||
| AK172470.1 | BAE43023.1 | |||
| AK173275.1 | BAD32553.1 | |||
| BC030930.1 | AAH30930.1 | |||
| BC092278.1 | AAH92278.1 |
More Information
Set Membership: |
|

Genomic Map