Assay Details
Target Gene Details
Entrez Gene ID: | 27413 |
Gene Name: | ATP-binding cassette, sub-family B (MDR/TAP), member 11 |
Gene Aliases: |
ABC16, Bsep, Lith1, PFIC2, PGY4, SPGP |
Location: |
Chr.2:69238282-69342842 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcb11 | NM_021022.3 | NP_066302.2 | ||
| AF133903.1 | AAF14372.1 | |||
| AK144553.1 |
More Information
Set Membership: |
|

Genomic Map