Assay Details
Target Gene Details
Entrez Gene ID: | 11636 |
Gene Name: | adenylate kinase 1 |
Gene Aliases: |
Ak-1, B430205N08Rik |
Location: |
Chr.2:32621758-32635058 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ak1 | NM_001198790.1 | NP_001185719.1 | ||
| NM_001198791.1 | NP_001185720.1 | |||
| NM_001198792.1 | NP_001185721.1 | |||
| NM_021515.3 | NP_067490.1 | |||
| AJ010108.1 | CAB52407.1 | |||
| AJ010109.1 | CAB52408.1 | |||
| AK046613.1 | BAC32808.1 | |||
| AK089270.1 | BAC40822.1 | |||
| BC014802.1 | AAH14802.1 | |||
| BC054366.1 | AAH54366.1 | |||
| CF950379.1 | ||||
| DQ486026.1 |
More Information
Set Membership: |
|

Genomic Map