Assay Details
Target Gene Details
Entrez Gene ID: | 228356 |
Gene Name: | RIKEN cDNA 1110051M20 gene |
Gene Aliases: |
AI586322 |
Location: |
Chr.2:91273240-91444713 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1110051M20Rik | BC089006.1 | 9 | 4277 | AAH89006.1 |
Target Gene Details
Entrez Gene ID: | 77038 |
Gene Name: | ADP-ribosylation factor GTPase activating protein 2 |
Gene Aliases: |
2310032E02Rik, Zfp289 |
Location: |
Chr.2:91265115-91277371 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arfgap2 | NM_001166024.1 | NP_001159496.1 | ||
| NM_023854.2 | NP_076343.2 | |||
| AF229439.1 | AAF91258.1 | |||
| AK009565.1 | BAB26362.1 | |||
| AK149837.1 | BAE29114.1 | |||
| AK168489.1 | BAE40376.1 | |||
| AK169144.1 | BAE40923.1 | |||
| BC005495.1 | AAH05495.1 |
More Information
Set Membership: |
|

Genomic Map