Assay Details
Target Gene Details
Entrez Gene ID: | 17222 |
Gene Name: | anaphase promoting complex subunit 1 |
Gene Aliases: |
2610021O03Rik, AI047775, AI853536, AW547281, Apc1, Mcpr, tsg24 |
Location: |
Chr.2:128610083-128687410 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 32 - Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Anapc1 | NM_008569.2 | NP_032595.2 | ||
| AK034858.1 | ||||
| AK090134.1 | ||||
| BC139002.1 | ||||
| BC145492.1 | ||||
| X80169.1 | CAA56450.1 |
More Information
Set Membership: |
|

Genomic Map