Assay Details
Target Gene Details
Entrez Gene ID: | 11602 |
Gene Name: | angiopoietin 4 |
Gene Aliases: |
Agpt4, Ang3 |
Location: |
Chr.2:151911332-151944654 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Angpt4 | NM_009641.1 | 2 | 420 | NP_033771.1 |
| AF113707.1 | 2 | 420 | AAD21586.1 | |
| BC129965.1 | 2 | 420 | ||
| BC129966.1 | 2 | 420 |
More Information
Set Membership: |
|

Genomic Map