Assay Details
Target Gene Details
Entrez Gene ID: | 77794 |
Gene Name: | ADAMTS-like 2 |
Gene Aliases: |
A930008K15Rik, tcp-1 |
Location: |
Chr.2:27079342-27108613 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adamtsl2 | NM_029981.1 | NP_084257.1 | ||
| AB096928.1 | BAC76875.1 | |||
| AK156991.1 | BAE33926.1 | |||
| BC128568.1 | ||||
| BC129984.1 |
More Information
Set Membership: |
|

Genomic Map