Assay Details
Target Gene Details
Entrez Gene ID: | 77794 |
Gene Name: | ADAMTS-like 2 |
Gene Aliases: |
A930008K15Rik, tcp-1 |
Location: |
Chr.2:27079342-27108613 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adamtsl2 | NM_029981.1 | 17 | 2671 | NP_084257.1 |
| AB096928.1 | 17 | 2671 | BAC76875.1 | |
| AK020831.1 | 1 | 133 | BAB32221.1 | |
| AK156991.1 | 17 | 2674 | BAE33926.1 | |
| AK172978.1 | 5 | 5077 | BAD32256.1 | |
| BC128568.1 | 17 | 2623 | ||
| BC129984.1 | 17 | 2623 |
More Information
Set Membership: |
|

Genomic Map