Assay Details
Target Gene Details
Entrez Gene ID: | 319196 |
Gene Name: | ankyrin repeat and EF-hand domain containing 1 |
Gene Aliases: |
AV276460, Ankrd5 |
Location: |
Chr.2:136532321-136555854 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankef1 | NM_175667.4 | NP_783598.1 | ||
| BC120825.1 |

Genomic Map