Assay Details
Target Gene Details
Entrez Gene ID: | 72323 |
Gene Name: | ankyrin repeat and SOCS box-containing 6 |
Gene Aliases: |
2510004M11Rik, AA409356 |
Location: |
Chr.2:30823097-30828300 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asb6 | NM_133346.2 | 4 | 569 | NP_579924.1 |
| AK010909.1 | 4 | 550 | ||
| AK049312.1 | 4 | 558 | ||
| AK088890.1 | 4 | 504 | BAC40635.1 | |
| AK190893.1 | 1 | 47 | ||
| AY665653.1 | 4 | 455 | AAT76177.1 | |
| BC019192.1 | 4 | 561 | AAH19192.1 |
More Information
Set Membership: |
|

Genomic Map