Assay Details
Target Gene Details
Entrez Gene ID: | 65964 |
Gene Name: | sterile alpha motif and leucine zipper containing kinase AZK |
Gene Aliases: |
AV006891, B230120H23Rik, MLTK, MLTKalpha, MLTKbeta |
Location: |
Chr.2:72285637-72442610 on Build GRCm38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zak | NM_023057.5 | NP_075544.1 | ||
| AB049731.1 | BAB16442.1 | |||
| AK132186.1 | BAE21021.1 | |||
| BC023718.1 | AAH23718.1 |
More Information
Set Membership: |
|

Genomic Map