Assay Details
Target Gene Details
Entrez Gene ID: | 11898 |
Gene Name: | argininosuccinate synthetase 1 |
Gene Aliases: |
AA408052, ASS, Ass-1, fold |
Location: |
Chr.2:31470270-31520670 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ass1 | NM_007494.3 | NP_031520.1 | ||
| AK146635.1 | BAE27321.1 | |||
| AK149489.1 | BAE28914.1 | |||
| BC002074.1 | AAH02074.1 | |||
| BC087556.1 | AAH87556.1 | |||
| M31690.1 | AAA37266.1 |
More Information
Set Membership: |
|

Genomic Map