Assay Details
Target Gene Details
Entrez Gene ID: | 227624 |
Gene Name: | RAB, member RAS oncogene family-like 6 |
Gene Aliases: |
B230208H17Rik, Rbel1, Rbel1a, Rbel1b, parf |
Location: |
Chr.2:25583018-25608446 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rabl6 | NM_001024616.1 | 10 | 1403 | NP_001019787.2 |
| AK045520.1 | 10 | 1425 | BAC32404.1 | |
| AK081440.1 | 10 | 1413 | BAC38220.1 | |
| AK157020.1 | 10 | 1438 | ||
| AK162461.1 | 6 | 783 | BAE36932.1 | |
| AK171643.1 | 10 | 1414 | BAE42583.1 | |
| AK215518.1 | 2 | 176 | ||
| AK219937.1 | 2 | 230 | ||
| BC023692.1 | 10 | 1431 | AAH23692.1 | |
| BC058585.1 | 10 | 1416 | AAH58585.1 | |
| BC085507.1 | 10 | 1403 |
More Information
Set Membership: |
|

Genomic Map