Assay Details
Target Gene Details
Entrez Gene ID: | 215095 |
Gene Name: | astacin-like metalloendopeptidase (M12 family) |
Gene Aliases: |
7420700F21, C87576, Sas1b |
Location: |
Chr.2:127338639-127357655 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Astl | NM_172539.3 | 1 | 74 | NP_766127.1 |
| AK033037.1 | 1 | 74 | BAC28134.1 |
More Information
Set Membership: |
|

Genomic Map