Assay Details
Target Gene Details
Entrez Gene ID: | 215095 |
Gene Name: | astacin-like metalloendopeptidase (M12 family) |
Gene Aliases: |
7420700F21, C87576, Sas1b |
Location: |
Chr.2:127338639-127357655 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Astl | NM_001291003.1 | NP_001277932.1 | ||
| NM_172539.3 | NP_766127.1 | |||
| AJ537599.2 | CAD61264.2 | |||
| AK033037.1 | BAC28134.1 | |||
| BC064729.1 | ||||
| DV649054.1 | ||||
| FJ187790.1 | ||||
| FJ187791.1 | ||||
| FJ187792.1 | ||||
| FJ187793.1 | ||||
| FJ187794.1 | ||||
| FJ187795.1 |
More Information
Set Membership: |
|

Genomic Map