Assay Details
Target Gene Details
Entrez Gene ID: | 227624 |
Gene Name: | RAB, member RAS oncogene family-like 6 |
Gene Aliases: |
B230208H17Rik, Rbel1, Rbel1a, Rbel1b, parf |
Location: |
Chr.2:25583018-25608446 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rabl6 | NM_001024616.1 | NP_001019787.2 | ||
| AK045520.1 | BAC32404.1 | |||
| AK081440.1 | BAC38220.1 | |||
| AK157020.1 | ||||
| AK162461.1 | BAE36932.1 | |||
| AK171643.1 | BAE42583.1 | |||
| AK204116.1 | ||||
| BC023692.1 | AAH23692.1 | |||
| BC058585.1 | AAH58585.1 | |||
| BC085507.1 |
More Information
Set Membership: |
|

Genomic Map