Assay Details
Target Gene Details
Entrez Gene ID: | 228850 |
Gene Name: | Ral GTPase activating protein, beta subunit (non-catalytic) |
Gene Aliases: |
9330195P16, AI507211, B230339M05Rik, mKIAA1219, p170 |
Location: |
Chr.2:158409829-158499253 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ralgapb | NM_001291137.1 | 6 | 1088 | NP_001278066.1 |
| NM_001291138.1 | 6 | 1088 | NP_001278067.1 | |
| AK046067.1 | 6 | 1063 | BAC32589.1 | |
| BC138377.1 | 6 | 990 |

Genomic Map