Assay Details
Target Gene Details
Entrez Gene ID: | 76117 |
Gene Name: | Rho GTPase activating protein 15 |
Gene Aliases: |
5830480G12Rik |
Location: |
Chr.2:43748783-44395953 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap15 | NM_153820.3 | NP_722542.2 | ||
| AK018058.1 | ||||
| AK036520.1 | ||||
| AK040796.1 | ||||
| AK170537.1 | BAE41865.1 | |||
| BC024887.1 | AAH24887.1 | |||
| BC034881.1 | AAH34881.1 |
Target Gene Details
Entrez Gene ID: | 320872 |
Gene Name: | Rho GTPase activating protein 15, opposite strand |
Gene Aliases: |
A430092G05Rik |
Location: |
Chr.2:44059238-44065324 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap15os | NR_040622.1 | |||
| AK078625.1 |
More Information
Set Membership: |
|

Genomic Map