Assay Details
Target Gene Details
Entrez Gene ID: | 11990 |
Gene Name: | attractin |
Gene Aliases: |
AW558010, Mgca, mKIAA0548, mahogany, mg |
Location: |
Chr.2:130906447-131030333 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 24 - Intron 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atrn | NM_009730.2 | NP_033860.2 | ||
| AB093245.1 | BAC41429.1 | |||
| AF116897.1 | AAD20947.1 | |||
| AF119821.1 | AAD25372.1 | |||
| AK155069.1 | ||||
| BC141054.1 | ||||
| CK780901.1 |
More Information
Set Membership: |
|

Genomic Map